missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobutane, 99%, stab. with copper

Product Code. 11349599
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
Unit Size:
250g
500g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11349599 500 g 500g
11489037 50 g 50g
11499037 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11349599 Supplier Thermo Scientific Alfa Aesar Supplier No. B20619.36
 Restricted Item
Estimated Shipment: 26-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 513-48-4 | C4H9I | 184.02 g/mol

It is used as a solvent organic synthesis and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a solvent organic synthesis and as pharmaceutical intermediate.

Solubility
Insoluble in water. Soluble in alcohol and diethyl ether.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-48-4
Molecular Formula C4H9I
Molecular Weight (g/mol) 184.02
MDL Number MFCD00001072
InChI Key IQRUSQUYPCHEKN-UHFFFAOYNA-N
Synonym sec-butyl iodide, butane, 2-iodo, 2-butyl iodide, sec-iodobutane, 2-jodbutan, 2-jodbutan czech, 2-iodanylbutane, 2-iodobutan, 2-iodo-butane, sec.-butyl iodide
PubChem CID 10559
IUPAC Name 2-iodobutane
SMILES CCC(C)I

Specifications

Melting Point -104°C
Density 1.597
Boiling Point 119°C to 120°C
Flash Point 21°C (70°F)
Odor Ether-like
Linear Formula CH3CH2CHICH3
Refractive Index 1.4991
Quantity 500 g
UN Number UN2390
Beilstein 1718777
Sensitivity Light sensitive
Merck Index 14,1574
Solubility Information Insoluble in water. Soluble in alcohol and diethyl ether.
Formula Weight 184.02
Percent Purity 99%
Chemical Name or Material 2-Iodobutane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.