missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobiphenyl, 98%

Product Code. 10170512 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10170512 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10170512 Supplier Thermo Scientific Acros Supplier No. 367020250
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2113-51-1 | C12H9I | 280.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2113-51-1
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 300°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C12H9I
Refractive Index 1.6600 to 1.662
MDL Number MFCD00039396
Quantity 25 g
Synonym 2-iodobiphenyl, 2-iodo-1,1'-biphenyl, o-iodobiphenyl, 1,1'-biphenyl, 2-iodo, o-phenyliodobenzene, 2-iodo-biphenyl, biphenyl, 2-iodo, 2-iodo-1-phenylbenzene, 1-iodo-2-phenyl-benzene, 1,1'-biphenyl, iodo
InChI Key QFUYDAGNUJWBSM-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)C2=CC=CC=C2I
IUPAC Name 1-iodo-2-phenylbenzene
Molecular Weight (g/mol) 280.1
PubChem CID 75025
Formula Weight 280.1
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Iodobiphenyl, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Iodobiphenyl
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.