missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodobenzyl Alcohol, TRC
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Specifications
Specifications
| Chemical Name or Material | 2-Iodobenzyl Alcohol |
| CAS | 5159-41-1 |
| Molecular Formula | C7H7OI |
| Quantity | 2.5 g |
| Synonym | 2-Iodophenylmethanol, o-Iodobenzyl Alcohol, 2-Iodobenzenemethanol, 1-Hydroxymethyl-2-iodobenzene, 2-(Hydroxymethyl)-1-iodobenzene, 2-(Hydroxymethyl)iodobenzene |
| InChI Formula | InChI=1S/C7H7IO/c8-7-4-2-1-3-6(7)5-9/h1-4,9H,5H2 |
| SMILES | OCc1ccccc1I |
| IUPAC Name | (2-iodophenyl)methanol |
| Molecular Weight (g/mol) | 234.03 |
| Formula Weight | 233.9542 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction