missing translation for 'onlineSavingsMsg'
Learn More

2-Iodobenzotrifluoride, 99%, Thermo Scientific™

Product Code. 10388460 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10388460 25 mL Glass bottle 25mL
10018501 5 mL Glass bottle 5mL
10570014 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10388460 Supplier Thermo Scientific Acros Supplier No. 241980250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Chemical Identifiers

CAS 444-29-1
Molecular Formula C7H4F3I
Molecular Weight (g/mol) 272.01
MDL Number MFCD00001041
InChI Key IGZGUYVVBABKOY-UHFFFAOYSA-N
Synonym 2-iodobenzotrifluoride, 1-iodo-2-trifluoromethyl benzene, o-iodobenzotrifluoride, benzene, 1-iodo-2-trifluoromethyl, 2-iodobenztrifluoride, o-trifluoromethyliodobenzene, 2-iodo-alpha,alpha,alpha-trifluorotoluene, iodobenzotrifluoride, 2-iodo-a,a,a-trifluorotoluene, 2-iodobenzotrifloride
PubChem CID 67957
IUPAC Name 1-iodo-2-(trifluoromethyl)benzene
SMILES C1=CC=C(C(=C1)C(F)(F)F)I

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Yellow
Density 1.8900g/mL
Boiling Point 197.0°C to 198.0°C
Flash Point 81°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CF3(C6H4)I
Refractive Index 1.5290 to 1.5310
Quantity 25 mL
Beilstein 05, III, 725
Specific Gravity 1.89
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most common organic solvents
Formula Weight 272.01
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Iodobenzotrifluoride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Iodobenzotrifluoride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.