Learn More
2-Iodoanisole, 99%
Description
2-Iodoanisole has been used in palladium/copper-catalyzed synthesis of o-(1-alkynyl)anisoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Iodoanisole has been used in palladium/copper-catalyzed synthesis of o-(1-alkynyl)anisoles.
Solubility
Miscible with alcohol, chloroform and diethyl ether. Insoluble in water.
Notes
Light Sensitive, store in dark. Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
Specifications
Specifications
| Density | 1.8 |
| Boiling Point | 240°C to 242°C |
| Flash Point | >110°C (230°F) |
| Refractive Index | 1.622 |
| Quantity | 500 g |
| Beilstein | 1860243 |
| Sensitivity | Light sensitive |
| Merck Index | 14,5028 |
| Solubility Information | Miscible with alcohol,chloroform and diethyl ether. Insoluble in water. |
| Formula Weight | 234.04 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.