missing translation for 'onlineSavingsMsg'
Learn More

2-Iodoanisole, 99%

Product Code. 11459422
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11459422 100 g 100g
11449422 25 g 25g
11469422 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459422 Supplier Thermo Scientific Alfa Aesar Supplier No. A10150.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 529-28-2 | C7H7IO | 234.036 g/mol

2-Iodoanisole has been used in palladium/copper-catalyzed synthesis of o-(1-alkynyl)anisoles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodoanisole has been used in palladium/copper-catalyzed synthesis of o-(1-alkynyl)anisoles.

Solubility
Miscible with alcohol, chloroform and diethyl ether. Insoluble in water.

Notes
Light Sensitive, store in dark. Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 529-28-2
Molecular Formula C7H7IO
Molecular Weight (g/mol) 234.036
MDL Number MFCD00001039
InChI Key DVQWNQBEUKXONL-UHFFFAOYSA-N
Synonym 2-iodoanisole, o-iodoanisole, benzene, 1-iodo-2-methoxy, iodoanisole, 2-methoxyiodobenzene, o-anisyl iodide, 2-iodophenol methyl ether, anisole, o-iodo, 2-iodophenyl methyl ether, 1-iodo-2-methoxy-benzene
PubChem CID 68257
ChEBI CHEBI:16355
IUPAC Name 1-iodo-2-methoxybenzene
SMILES COC1=CC=CC=C1I

Specifications

Density 1.8
Boiling Point 240°C to 242°C
Flash Point >110°C (230°F)
Refractive Index 1.622
Quantity 100 g
Beilstein 1860243
Sensitivity Light sensitive
Merck Index 14,5028
Solubility Information Miscible with alcohol,chloroform and diethyl ether. Insoluble in water.
Formula Weight 234.04
Percent Purity 99%
Chemical Name or Material 2-Iodoanisole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.