missing translation for 'onlineSavingsMsg'
Learn More

2'-Iodoacetophenone, 99+%, Thermo Scientific Chemicals

Product Code. 10167093 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10167093 1 g Glass bottle 1g
10784194 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10167093 Supplier Thermo Scientific Acros Supplier No. 393130010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2142-70-3
Molecular Formula C8H7IO
Molecular Weight (g/mol) 246.04
MDL Number MFCD00094998
InChI Key XDXCBCXNCQGZPG-UHFFFAOYSA-N
Synonym 2'-iodoacetophenone, 1-2-iodophenyl ethanone, o-iodoacetophenone, 1-2-iodophenyl ethan-1-one, o-iodobenzoyl, acetophenone, o-iodo, acetophenone, 2'-iodo, ethanone, 1-iodophenyl, ethanone, 1-2-iodophenyl, 1-acetyl-2-iodobenzene
PubChem CID 240431
IUPAC Name 1-(2-iodophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1I

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color Yellow
Density 1.7460g/mL
Boiling Point 139°C to 140°C (12.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula IC6H4C(O)CH3
Refractive Index 1.617 to 1.619 (20°C, 589nm)
Quantity 1 g
Specific Gravity 1.746
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in most organic solvents
Formula Weight 246.04
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2'-Iodoacetophenone
Show More Show Less
compliance-icons
Product Identifier
  • 2'-Iodoacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.