missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-p-xylene, 98+%

Product Code. 11334434
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11334434 50 g 50g
11324434 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11334434 Supplier Thermo Scientific Alfa Aesar Supplier No. L01055.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1122-42-5 | C8H9I | 232.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-42-5
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00013708
InChI Key WYZVNUSNUCABRF-UHFFFAOYSA-N
Synonym 2-iodo-p-xylene, 1,4-dimethyl-2-iodobenzene, p-xylene, 2-iodo, 1-iodo-2,5-dimethylbenzene, benzene, 2-iodo-1,4-dimethyl, 2-iodo-4-xylene, 2,5-dimethyliodobenzene, 2-iodo-1,4-dimethyl-benzene, 2,5-dimethyl-1-iodobenzene, pubchem3875
PubChem CID 70731
IUPAC Name 2-iodo-1,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)C)I

Specifications

Density 1.617
Quantity 50 g
Refractive Index 1.598
Beilstein 1905728
Boiling Point 229°C to 230°C
Formula Weight 232.07
Percent Purity ≥98%
Chemical Name or Material 2-Iodo-p-xylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.