missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-m-xylene, 98%

Product Code. 11495483
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11495483 5 g 5g
11405493 25 g 25g
11415493 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495483 Supplier Thermo Scientific Alfa Aesar Supplier No. A13439.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 608-28-6 | C8H9I | 232.064 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 608-28-6
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.064
MDL Number MFCD00019012
InChI Key QTUGGVBKWIYQSS-UHFFFAOYSA-N
Synonym 2-iodo-m-xylene, 2,6-dimethyliodobenzene, m-xylene, 2-iodo, benzene, 2-iodo-1,3-dimethyl, 1,3-dimethyl-2-iodobenzene, 2-iodo-1,3-dimethyl-benzene, 2,6-dimethyl-1-iodobenzene, 2-iodo-o-xylene, pubchem3867, acmc-1axwc
PubChem CID 69097
IUPAC Name 2-iodo-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)I

Specifications

Density 1.62
Quantity 5 g
Refractive Index 1.6025
Beilstein 1857414
Boiling Point 230°C to 231°C
Formula Weight 232.07
Percent Purity 98%
Chemical Name or Material 2-Iodo-m-xylene
compliance-icons
Product Identifier
  • 2,6-Dimethyliodobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-iodo-1,3-dimethylbenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.