missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo Adenosine35109-88-7, TRC

Product Code. 30401590
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30401590 500 mg 500mg
30401289 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30401590 Supplier Toronto Research Chemicals Supplier No. TRCI686000500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Iodo Adenosine
CAS 35109-88-7
Molecular Formula C10 H12 I N5 O4
Quantity 500 mg
InChI Formula InChI=1S/C10H12IN5O4/c11-10-14-7(12)4-8(15-10)16(2-13-4)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H2,12,14,15)/t3-,5-,6-,9-/m1/s1
SMILES Nc1nc(I)nc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O
IUPAC Name (2R,3R,4S,5R)-2-(6-amino-2-iodopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol) 393.14
Formula Weight 392.9934
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.