missing translation for 'onlineSavingsMsg'
Learn More

(2-Iodo-5-methoxyphenyl)boronic Acid, TRC

Product Code. 30407557
Change view
Click to view available options
Quantity:
1 g
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30407557 1 g -
30389913 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407557 Supplier Toronto Research Chemicals Supplier No. TRCI7084801G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89694-50-8
Molecular Formula C7 H8 B I O3
Molecular Weight (g/mol) 277.85
Synonym B-(2-Iodo-5-methoxyphenyl)-boronic Acid, 2-Iodo-5-methoxybenzeneboronic Acid
IUPAC Name (2-iodo-5-methoxyphenyl)boronic acid
SMILES COc1ccc(I)c(c1)B(O)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C7H8BIO3/c1-12-5-2-3-7(9)6(4-5)8(10)11/h2-4,10-11H,1H3
Formula Weight 277.9611
Chemical Name or Material (2-Iodo-5-methoxyphenyl)boronic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.