missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-5’-ethylcarboxamido Adenosine, TRC

Product Code. 30396305
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396305 50 mg 50mg
30403884 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396305 Supplier Toronto Research Chemicals Supplier No. TRCI70600050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141018-29-3
Molecular Formula C13 H17 I N6 O5
Molecular Weight (g/mol) 464.22
Synonym 2-Iodo-5-Ethylcarboxamido Adenosine
IUPAC Name [(2R,3S,4R,5R)-5-(6-amino-2-iodopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methyl N-ethylcarbamate
SMILES CCNC(=O)OC[C@H]1O[C@H]([C@H](O)[C@@H]1O)n2cnc3c(N)nc(I)nc23

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H17IN6O5/c1-2-16-13(23)24-3-5-7(21)8(22)11(25-5)20-4-17-6-9(15)18-12(14)19-10(6)20/h4-5,7-8,11,21-22H,2-3H2,1H3,(H,16,23)(H2,15,18,19)/t5-,7-,8-,11-/m1/s1
Formula Weight 464.0305
Chemical Name or Material 2-Iodo-5'-ethylcarboxamido Adenosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.