missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-4-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™

Product Code. p-7031682
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455607 250 mg 250mg
15435607 1 g 1g
15445607 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15455607 Supplier Thermo Scientific Alfa Aesar Supplier No. H32839.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
The 10-chloro-8-fluoro-10, 11-dihydro-2-trifluoromethyldibenzothiepin used as the starting material can be prepared starting from 2-iodo--4-trifluoromethyl-benzoic acid and 4-fluoro-thiophenol.

Solubility
Slightly soluble.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54507-44-7
Molecular Formula C8H4F3IO2
Molecular Weight (g/mol) 316.018
MDL Number MFCD06408817
InChI Key WCLYLTXCKMZNOH-UHFFFAOYSA-N
Synonym 2-iodo-4-trifluoromethyl benzoic acid, 2-iodo-4-trifluoromethyl-benzoic acid, benzoic acid, 2-iodo-4-trifluoromethyl, 2-iodo-4-trifluoromethylbenzoicacid, 2-iodo-4-trifluoromethyl-benzoicacid
PubChem CID 16244090
IUPAC Name 2-iodo-4-(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C=C1C(F)(F)F)I)C(=O)O

Specifications

Quantity 250 mg
Sensitivity Light sensitive
Solubility Information Slightly soluble.
Formula Weight 316.02
Percent Purity 98%
Chemical Name or Material 2-Iodo-4-(trifluoromethyl)benzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.