missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-4-nitrotoluene, 97%

Product Code. 11487034
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487034 25 g 25g
11477034 5 g 5g
11497034 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487034 Supplier Thermo Scientific Alfa Aesar Supplier No. B21491.14
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7745-92-8 | C7H6INO2 | 263.03 g/mol

2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodo-4-nitrotoluene is used as a pharmaceutical intermediate, chemical synthesis intermediate.

Solubility
Soluble in toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7745-92-8
Molecular Formula C7H6INO2
Molecular Weight (g/mol) 263.03
MDL Number MFCD00024329
InChI Key BUQSRXQJUZTIEW-UHFFFAOYSA-N
Synonym 2-iodo-4-nitrotoluene, benzene, 2-iodo-1-methyl-4-nitro, 2-iodo-1-methyl-4-nitro-benzene, pubchem3957, acmc-209pa2, toluene, 2-iodo-4-nitro, timtec-bb sbb005821
PubChem CID 82188
IUPAC Name 2-iodo-1-methyl-4-nitrobenzene
SMILES CC1=CC=C(C=C1I)[N+]([O-])=O

Specifications

Quantity 25 g
Sensitivity Light sensitive
Solubility Information Soluble in toluene.
Formula Weight 263.03
Percent Purity 97%
Chemical Name or Material 2-Iodo-4-nitrotoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.