missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-4,6-dimethylaniline, 98%, Thermo Scientific Chemicals

Product Code. 15498367
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498367 1 g 1g
15408377 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498367 Supplier Thermo Scientific Alfa Aesar Supplier No. H51875.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4102-54-9 | Molecular Formula: C8H10IN | Molecular Weight: 247.079 g/mol

It is employed in the synthesis of benzothiazoles through copper-catalyzed one-pot three-component reactions with use of sodium hydrosulfide as a sulfur surrogate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of benzothiazoles through copper-catalyzed one-pot three-component reactions with use of sodium hydrosulfide as a sulfur surrogate.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from light and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store in dark.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4102-54-9
Molecular Formula C8H10IN
Molecular Weight (g/mol) 247.079
MDL Number MFCD07779005
InChI Key ODPOIEACUVQCBZ-UHFFFAOYSA-N
Synonym 2,4-dimethyl-6-iodoaniline, 2-iodo-4,6-di-methylaniline, benzenamine,2-iodo-4,6-dimethyl
PubChem CID 14040290
IUPAC Name 2-iodo-4,6-dimethylaniline
SMILES CC1=CC(=C(C(=C1)I)N)C

Specifications

Melting Point 64°C to 65°C
Quantity 1 g
UN Number UN2811
Solubility Information Insoluble in water.
Formula Weight 247.08
Percent Purity 98%
Chemical Name or Material 2-Iodo-4,6-dimethylaniline
compliance-icons
Product Identifier
  • 2-Iodo-4,6-dimethylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Iodo-4,6-dimethylaniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.