missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-3-methylbenzoic acid, 98%

Product Code. 11389885
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11389885 5 g 5g
11379885 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11389885 Supplier Thermo Scientific Alfa Aesar Supplier No. L19876.06
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 108078-14-4 | C8H7IO2 | 262.046 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108078-14-4
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.046
MDL Number MFCD00079764
InChI Key PZUXUOZSSYKAMX-UHFFFAOYSA-N
Synonym 2-iodo-m-toluic acid, benzoic acid,2-iodo-3-methyl, 2-iodo-3-methyl-benzoic acid, pubchem12741, acmc-1bsuz, 3-methyl-2-iodobenzoic acid, 2-iodo-3-methyl benzoic acid, buttpark 148\07-11, 2-iodo-3-methylbenzoic acid
PubChem CID 2759365
IUPAC Name 2-iodo-3-methylbenzoic acid
SMILES CC1=CC=CC(=C1I)C(=O)O

Specifications

Melting Point 150°C to 153°C
Quantity 5 g
Beilstein 2439521
Sensitivity Light sensitive
Formula Weight 262.05
Percent Purity 98%
Chemical Name or Material 2-Iodo-3-methylbenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.