missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxythiophenol, 97%, Thermo Scientific Chemicals

Product Code. 11363206
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11363206 5 g 5g
11353206 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11363206 Supplier Thermo Scientific Alfa Aesar Supplier No. L12519.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1121-24-0 | Molecular Formula: C6H6OS | Molecular Weight: 126.173 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-24-0
Molecular Formula C6H6OS
Molecular Weight (g/mol) 126.173
MDL Number MFCD00040447
InChI Key VMKYTRPNOVFCGZ-UHFFFAOYSA-N
Synonym 2-hydroxythiophenol, 2-mercaptophenol, o-mercaptophenol, phenol, 2-mercapto, 2-hydroxy thiophenol, 2-hydroxybenzenethiol, o-hydroxythiophenol, o-hydroxybenzenothiol, 2-hydroxybenzenothiol, mercaptophenol
PubChem CID 70718
IUPAC Name 2-sulfanylphenol
SMILES C1=CC=C(C(=C1)O)S

Specifications

Melting Point 19°C
Density 1.22
Boiling Point 217°C
Flash Point 97°C (206°F)
Odor Unpleasant
Refractive Index 1.614
Quantity 5 g
UN Number UN2922
Beilstein 1280884
Sensitivity Air Sensitive
Formula Weight 126.18
Percent Purity 97%
Chemical Name or Material 2-Hydroxythiophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.