missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxysuccinic Acid Methyl Ester (>90%), TRC

Product Code. 30396859
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396859 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396859 Supplier Toronto Research Chemicals Supplier No. TRCH9535605G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxysuccinic Acid Methyl Ester
CAS 140235-34-3
Molecular Formula C5 H8 O5
Quantity 5 g
Synonym Butanedioic acid, 2-hydroxy-, 1-methyl ester, Butanedioic acid, hydroxy-, 1-methyl ester (9CI), 1-Methyl malate, 2-Hydroxysuccinic Acid Methyl Ester, 3-Hydroxy-4-methoxy-4-oxobutanoic acid, Malic acid 1-methyl ester, 1-Methyl 2-hydroxybutanedioate
InChI Formula InChI=1S/C5H8O5/c1-10-5(9)3(6)2-4(7)8/h3,6H,2H2,1H3,(H,7,8)
SMILES COC(=O)C(O)CC(=O)O
IUPAC Name 3-hydroxy-4-methoxy-4-oxobutanoic acid
Molecular Weight (g/mol) 148.11
Formula Weight 148.0372
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.