Learn More
2-Hydroxysaclofen, Thermo Scientific™
Selective antagonist at GABA-B receptors
£259.00 - £881.00
Chemical Identifiers
| CAS | 117354-64-0 |
|---|---|
| Molecular Formula | C9H12ClNO4S |
| Molecular Weight (g/mol) | 265.708 |
| MDL Number | MFCD00069282 |
| InChI Key | WBSMZVIMANOCNX-UHFFFAOYSA-N |
| Synonym | 2-hydroxysaclofen, 2-oh-saclofen, 2-hydroxy-saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropane-1-sulfonic acid, 2-hydroxy-s---saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropylsulfonic acid, 3-amino-2-4-chlorophenyl-2-hydroxypropanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxybenzeneethanesulfonic acid, benzeneethanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxy, 2-hydroxysaclofen, solid |
| PubChem CID | 1564 |
| IUPAC Name | 3-amino-2-(4-chlorophenyl)-2-hydroxypropane-1-sulfonic acid |
| SMILES | C1=CC(=CC=C1C(CN)(CS(=O)(=O)O)O)Cl |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15415789
|
Thermo Scientific Alfa Aesar
J63588.MA |
10mg |
£259.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15425789
|
Thermo Scientific Alfa Aesar
J63588.MF |
50mg |
£881.00
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 117354-64-0 | |
| 265.708 | |
| WBSMZVIMANOCNX-UHFFFAOYSA-N | |
| 1564 | |
| C1=CC(=CC=C1C(CN)(CS(=O)(=O)O)O)Cl |
| C9H12ClNO4S | |
| MFCD00069282 | |
| 2-hydroxysaclofen, 2-oh-saclofen, 2-hydroxy-saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropane-1-sulfonic acid, 2-hydroxy-s---saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropylsulfonic acid, 3-amino-2-4-chlorophenyl-2-hydroxypropanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxybenzeneethanesulfonic acid, benzeneethanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxy, 2-hydroxysaclofen, solid | |
| 3-amino-2-(4-chlorophenyl)-2-hydroxypropane-1-sulfonic acid |
Specifications
| 117354-64-0 | |
| MFCD00069282 | |
| 2-hydroxysaclofen, 2-oh-saclofen, 2-hydroxy-saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropane-1-sulfonic acid, 2-hydroxy-s---saclofen, 3-amino-2-4-chlorophenyl-2-hydroxypropylsulfonic acid, 3-amino-2-4-chlorophenyl-2-hydroxypropanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxybenzeneethanesulfonic acid, benzeneethanesulfonic acid, beta-aminomethyl-4-chloro-beta-hydroxy, 2-hydroxysaclofen, solid | |
| WBSMZVIMANOCNX-UHFFFAOYSA-N | |
| 3-amino-2-(4-chlorophenyl)-2-hydroxypropane-1-sulfonic acid | |
| 1564 | |
| 2-Hydroxysaclofen |
| C9H12ClNO4S | |
| UN1759 | |
| Soluble in water at 1mg/ml; Also soluble in DMSO,0;1M HCl and methanol | |
| C1=CC(=CC=C1C(CN)(CS(=O)(=O)O)O)Cl | |
| 265.708 | |
| 265.71 |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
missing translation for 'rtecsNumber' : DA0359200
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.