missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyquinoxaline, 98%, Thermo Scientific™

Product Code. 15269936 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15269936 25 g 25g
15279936 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15269936 Supplier Thermo Scientific Acros Supplier No. 454460250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1196-57-2
Molecular Formula C8H6N2O
Molecular Weight (g/mol) 146.15
MDL Number MFCD00006722
InChI Key FFRYUAVNPBUEIC-UHFFFAOYSA-N
Synonym 2-hydroxyquinoxaline, quinoxalin-2-ol, 2-quinoxalinol, 2 1h-quinoxalinone, quinoxalin-2 1h-one, quinoxalin-2-one, 2-quinoxalinone, 3-quinoxalinone, 2-quinoxalone, 1,2-dihydroquinoxalin-2-one
PubChem CID 14526
ChEBI CHEBI:38890
IUPAC Name 1H-quinoxalin-2-one
SMILES O=C1NC2=CC=CC=C2N=C1

Specifications

Melting Point 271.0°C to 272.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: soluble in hot methanol
Formula Weight 146.15
Percent Purity 98%
Chemical Name or Material 2-Hydroxyquinoxaline
compliance-icons
Product Identifier
  • 2-Hydroxyquinoxaline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.