missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyquinoline, 98%

Product Code. 10187750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10187750 25 g Glass bottle 25g
10591124 1 g Glass bottle 1g
10463361 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10187750 Supplier Thermo Scientific Acros Supplier No. 418810250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 59-31-4 | C9H7NO | 145.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-31-4
Molecular Formula C9H7NO
Molecular Weight (g/mol) 145.16
MDL Number MFCD00006743
InChI Key LISFMEBWQUVKPJ-UHFFFAOYSA-N
Synonym 2-hydroxyquinoline, 2-quinolinol, quinolin-2-ol, carbostyril, 2 1h-quinolinone, 2-quinolone, quinolin-2 1h-one, quinolinol, quinolinone, alpha-quinolone
PubChem CID 6038
ChEBI CHEBI:18289
IUPAC Name 1H-quinolin-2-one
SMILES C1=CC=C2C(=C1)C=CC(=O)N2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 197.0°C to 200.0°C
Color Brown-Purple or Purple to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 1826
Formula Weight 145.16
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Hydroxyquinoline, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Hydroxyquinoline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.