missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyrimidine Hydrochloride, TRC

Product Code. 30396728
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396728 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30396728 Supplier Toronto Research Chemicals Supplier No. TRCH9530405G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxypyrimidine Hydrochloride
CAS 38353-09-2
Molecular Formula C4H4N2O .HCl
Quantity 5 g
Synonym 2(1H)-Pyrimidinone, 1,2-Dihydropyrimidin-2-one Hydrochloride, 1H-Pyrimidin-2-one Hydrochloride, 2(1H)-Pyrimidone, 2-Hydroxypyrimidine, 2-Oxopyrimidine Hydrochloride, 2-Pyrimidinol Hydrochloride, 2-Pyrimidinone Hydrochloride, 2-Pyrimidone Hydrochloride, NSC 618280 Hydrochloride
InChI Formula InChI=1S/C4H4N2O.ClH/c7-4-5-2-1-3-6-4;/h1-3H,(H,5,6,7);1H
SMILES O=C1N=CC=CN1.Cl
IUPAC Name pyrimidin-2(1H)-one hydrochloride
Molecular Weight (g/mol) 132.55
Formula Weight 132.49
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.