missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyrimidine hydrochloride, 98+%

Product Code. 11444754
Change view
Click to view available options
Quantity:
25 g
500 g
Unit Size:
25g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444754 500 g 500g
11424754 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11444754 Supplier Thermo Scientific Alfa Aesar Supplier No. A13732.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 38353-09-2 | C4H5ClN2O | 132.547 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38353-09-2
Molecular Formula C4H5ClN2O
Molecular Weight (g/mol) 132.547
MDL Number MFCD00012781
InChI Key IAJINJSFYTZPEJ-UHFFFAOYSA-N
Synonym 2-hydroxypyrimidine hydrochloride, pyrimidin-2-ol hydrochloride, 2 1h-pyrimidinone, monohydrochloride, 2-hydroxypyrimidine hcl, 2-pyrimidinone hydrochloride, pyrimidin-2 1h-one hydrochloride, 2-hydroxypyrimidinehydrochloride, pyrimidin-2-one monohydrochloride, 1,2-dihydropyrimidin-2-one hydrochloride, pyrimidin-2-one hydrochloride
PubChem CID 122774
IUPAC Name 1H-pyrimidin-2-one;hydrochloride
SMILES C1=CNC(=O)N=C1.Cl

Specifications

Melting Point ∼210°C (decomposition)
Quantity 500 g
Beilstein 3560963
Sensitivity Hygroscopic
Formula Weight 132.55
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxypyrimidine hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.