missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyrimidine-5-boronic acid pinacol ester, 95%

Product Code. 15427645
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15427645

Brand: Thermo Scientific Alfa Aesar H26976.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1073354-84-3 | C10H15BN2O3 | 222.051 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1073354-84-3
Molecular Formula C10H15BN2O3
Molecular Weight (g/mol) 222.051
MDL Number MFCD09037497
InChI Key JRCSRFGCQSLWNZ-UHFFFAOYSA-N
Synonym 2-hydroxypyrimidine-5-boronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-ol, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidin-2-ol, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-ol, 2-hydroxypyrimidin-5-ylboronic acid pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyrimidinol, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2 1h-one
PubChem CID 44755196
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrimidin-2-one
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CNC(=O)N=C2

Specifications

Melting Point 201°C to 204°C
Quantity 250 mg
Formula Weight 222.05
Percent Purity 95%
Chemical Name or Material 2-Hydroxypyrimidine-5-boronic acid pinacol ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.