missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyridine, 97%

Product Code. 10215450 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
50 g
250 g
Packaging:
Glass bottle
Plastic bottle
Unit Size:
250g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215450 5 g Glass bottle 5g
10030940 50 g Plastic bottle 50g
10358983 250 g Plastic bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10215450 Supplier Thermo Scientific Acros Supplier No. 121840050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142-08-5 | C5H5NO | 95.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142-08-5
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.10
MDL Number MFCD00006268
InChI Key UBQKCCHYAOITMY-UHFFFAOYSA-N
Synonym 2-hydroxypyridine, pyridin-2-ol, 2-pyridinol, 2-pyridone, pyridin-2 1h-one, 2 1h-pyridinone, 2-pyridinone, 2-oxopyridine, 2 1h-pyridone, alpha-pyridone
PubChem CID 8871
ChEBI CHEBI:16540
IUPAC Name 1H-pyridin-2-one
SMILES O=C1NC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 102.0°C to 109.0°C
Color Brown to Colorless
Boiling Point 280.0°C to 281.0°C
Flash Point 210°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 21, 43
Fieser 03,157
Solubility Information Solubility in water: soluble in water. Other solubilities: slightly soluble in chloroform and alcohol, practically insoluble in ether and benzene
Formula Weight 95.1
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Hydroxypyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxypyridine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.