missing translation for 'onlineSavingsMsg'
Learn More

2-hydroxypyridine-5-boronic acid, 97%, Thermo Scientific™

Product Code. 10516534
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10516534 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10516534 Supplier Thermo Scientific Maybridge Supplier No. MAY00026DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 1054483-78-1
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C11H16BNO3
MDL Number MFCD06795671
Quantity 1 g
Synonym 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ol, 6-hydroxypyridine-3-boronic acid pinacol ester, 2-hydroxypyridine-5-boronic acid pinacol ester, 2-hydroxypyridine-5-boronic acid, pinacol ester, 5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-ol, 6-hydroxypyridine-3-boronic acid, pinacol ester, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2 1h-one, 2-hydroxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, amtb145, pyridin-2-ol-5-boronic acid pinacol ester
InChI Key WAUWXCUPDOXYKS-UHFFFAOYSA-N
SMILES CC1(C)OB(OC1(C)C)C1=CN=C(O)C=C1
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyridin-2-one
Molecular Weight (g/mol) 221.06
PubChem CID 17750253
Formula Weight 221.06
Percent Purity 97%
Chemical Name or Material 2-Hydroxypyridine-5-boronic acid, pinacol ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.