missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypyrazine, TRC

Product Code. 30398636
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30398636 100 g 100g
30406396 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30398636 Supplier Toronto Research Chemicals Supplier No. TRCH952710100G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxypyrazine
CAS 6270-63-9
Molecular Formula C4 H4 N2 O
Quantity 100 g
Synonym Pyrazinol (6CI,7CI,8CI), 1H-Pyrazin-2-one, 2-Oxo-1H-pyrazine, NSC 36081, Pyrazin-2(1H)-one, Pyrazin-2-ol, 2(1H)-Pyrazinone
InChI Formula InChI=1S/C4H4N2O/c7-4-3-5-1-2-6-4/h1-3H,(H,6,7)
SMILES O=C1NC=CN=C1
IUPAC Name 1H-pyrazin-2-one
Molecular Weight (g/mol) 96.087
Formula Weight 96.0324
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.