missing translation for 'onlineSavingsMsg'
Learn More

(2-Hydroxypropyl)propyl S-Ethyl Ester Carbamothioic Acid, TRC

Product Code. 30402577
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402577 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30402577 Supplier Toronto Research Chemicals Supplier No. TRCH65109510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material EPTC-2-hydroxypropyl
CAS 65109-69-5
Molecular Formula C9 H19 N O2 S
Quantity 10 mg
Synonym Carbamothioic acid, (2-hydroxypropyl)propyl-, S-ethyl ester, EPTC-2-hydroxypropyl
InChI Formula InChI=1S/C9H19NO2S/c1-4-6-10(7-8(3)11)9(12)13-5-2/h8,11H,4-7H2,1-3H3
SMILES CCCN(CC(C)O)C(=O)SCC
IUPAC Name S-ethyl N-(2-hydroxypropyl)-N-propylcarbamothioate
Molecular Weight (g/mol) 205.32
Formula Weight 205.1136
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.