missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxypropyl Hexadecanoate, TRC
Specifications
Specifications
| Chemical Name or Material | 1-Propylene Glycol Monopalmitate |
| CAS | 3539-33-1 |
| Molecular Formula | C19 H38 O3 |
| Quantity | 25 mg |
| Synonym | 2-Hydroxypropyl Hexadecanoate, Hexadecanoic Acid 2-Hydroxypropyl Ester, Palmitic acid, 2-hydroxypropyl ester (7CI,8CI), 1-Palmitoyl-1,2-propanediol, 1-Propylene glycol monopalmitate, 2-Hydroxypropyl hexadecanoate, Ceraphyl 28 |
| InChI Formula | InChI=1S/C19H38O3/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-19(21)22-17-18(2)20/h18,20H,3-17H2,1-2H3 |
| SMILES | CCCCCCCCCCCCCCCC(=O)OCC(C)O |
| IUPAC Name | 2-hydroxypropyl hexadecanoate |
| Molecular Weight (g/mol) | 314.503 |
| Formula Weight | 314.282 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?