missing translation for 'onlineSavingsMsg'
Learn More

(2-Hydroxypropyl)-β-cyclodextrin, MedChemExpress

Product Code. 16495987
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
Product Code. Quantity unitSize
16495987 5 g 5g
16440729 1 g 1g
16463268 10 g 10g
3 options
This item is not returnable. View return policy

Product Code. 16495987

Brand: Medchem Express HY101103

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (2-Hydroxypropyl)-β-cyclodextrin is a widely used drug delivery vehicle to improve the stability and bioavailability.

(2-Hydroxypropyl)-β-cyclodextrin is a widely used drug delivery vehicle to improve the stability and bioavailability.

Specifications

Chemical Name or Material (2-Hydroxypropyl)-β-cyclodextrin
CAS 128446-35-5
Color White
Purity Grade Notes Research
Quantity 5 g
Synonym Hydroxypropyl betadex Hydroxypropyl-β-cyclodextrin HP-β-CD
Solubility Information DMSO : 50 mg/mL (Need ultrasonic) ∣H2O : 50 mg/mL (Need ultrasonic)
SMILES CO[C@@H]1[C@H](O[R])[C@@H](O)[C@H](C)[C@@H](CO[R])O1.[C;D2]CC(O)C.[R].[7].[R=H or]
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.