missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypropyl Acrylate, TRC

Product Code. 30389871
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389871 50 mg 50mg
30403668 1 g -
30395493 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30389871 Supplier Toronto Research Chemicals Supplier No. TRCH99994050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 999-61-1
Molecular Formula C6 H10 O3
Molecular Weight (g/mol) 130.142
Synonym 2-Propenoic acid, 2-hydroxypropyl ester, Acrylic acid, 2-hydroxypropyl ester (6CI,8CI), 1,2-Propanediol, 1-acrylate (8CI), 2-Hydroxy-2-methylethyl acrylate, 2-Hydroxypropyl acrylate, 2HPA, Bisomer HPA, HOP-A, HPA, Kayarad HPA, Light Ester HOP-A, Light Ester HOP-A(N), β-Hydroxypropyl acrylate
IUPAC Name 2-hydroxypropyl prop-2-enoate
SMILES CC(O)COC(=O)C=C

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C6H10O3/c1-3-6(8)9-4-5(2)7/h3,5,7H,1,4H2,2H3
Formula Weight 130.063
Chemical Name or Material 2-Propenoic acid-2-hydroxypropyl ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.