missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyphenoxyacetic acid, 98+%

Product Code. 11336706
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336706 5 g 5g
11346706 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336706 Supplier Thermo Scientific Alfa Aesar Supplier No. L03897.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 6324-11-4 | C8H8O4 | 168.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6324-11-4
MDL Number MFCD00015454

Specifications

Melting Point 125°C to 130°C
Molecular Formula C8H8O4
Quantity 5 g
Beilstein 1948566
Synonym 2-hydroxyphenoxyacetic acid, 2-hydroxyphenoxy acetic acid, 2-2-hydroxyphenoxy acetic acid, o-hydroxyphenoxy acetic acid, acetic acid, hydroxyphenoxy, o-hydroxyphenoxyacetic acid, enamine_004711, 2-carboxymethoxy phenol, cambridge id 5156176
InChI Key LPXHPAYRONCQIF-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)O)OCC(=O)O
IUPAC Name 2-(2-hydroxyphenoxy)acetic acid
Molecular Weight (g/mol) 168.15
PubChem CID 22783
Formula Weight 168.15
Percent Purity ≥98%
Chemical Name or Material 2-Hydroxyphenoxyacetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.