missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxypentanoic Acid, TRC

Product Code. 30403715
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30403715 100 mg -
30402423 500 mg -
30394586 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403715 Supplier Toronto Research Chemicals Supplier No. TRCH250210100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 617-31-2
Molecular Formula C5 H10 O3
Molecular Weight (g/mol) 118.131
Synonym 2-Hydroxypentanoic Acid, (±)-2-Hydroxy-n-valeric Acid, (±)-2-Hydroxypentanoic Acid, (±)-2-Hydroxyvaleric Acid, (±)-alpha-Hydroxyvaleric Acid, 2-Hydroxyvaleric Acid, DL-2-Hydroxypentanoic Acid, DL-2-Hydroxyvaleric Acid, DL-alpha-Hydroxyvaleric Acid, NSC 67957, alpha-Hydroxy-n-valeric Acid, alpha-Hydroxypentanoic Acid, alpha-Hydroxyvaleric Acid
IUPAC Name 2-hydroxypentanoic acid
SMILES CCCC(O)C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C5H10O3/c1-2-3-4(6)5(7)8/h4,6H,2-3H2,1H3,(H,7,8)
Formula Weight 118.063
Chemical Name or Material 2-Hydroxypentanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.