missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxynicotinic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-146552
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
609-71-2
C6H5NO3
139.11
MFCD00010100
UEYQJQVBUVAELZ-UHFFFAOYSA-N
2-hydroxynicotinic acid, 2-hydroxypyridine-3-carboxylic acid, 2-oxo-1,2-dihydropyridine-3-carboxylic acid, 2-hydroxynicotinicacid, unii-tmq68q9ba3, 3-pyridinecarboxylic acid, 1,2-dihydro-2-oxo, 1,2-dihydro-2-oxonicotinic acid, tmq68q9ba3, 2-hydroxy-3-pyridinecarboxylic acid, 1,2-dihydro-2-oxo-3-pyridinecarboxylic acid
69114
2-oxo-1H-pyridine-3-carboxylic acid
OC(=O)C1=CC=CNC1=O
This item is not returnable. View return policy
609-71-2
C6H5NO3
139.11
MFCD00010100
UEYQJQVBUVAELZ-UHFFFAOYSA-N
2-hydroxynicotinic acid, 2-hydroxypyridine-3-carboxylic acid, 2-oxo-1,2-dihydropyridine-3-carboxylic acid, 2-hydroxynicotinicacid, unii-tmq68q9ba3, 3-pyridinecarboxylic acid, 1,2-dihydro-2-oxo, 1,2-dihydro-2-oxonicotinic acid, tmq68q9ba3, 2-hydroxy-3-pyridinecarboxylic acid, 1,2-dihydro-2-oxo-3-pyridinecarboxylic acid
69114
2-oxo-1H-pyridine-3-carboxylic acid
OC(=O)C1=CC=CNC1=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 259.0°C to 261.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 22, 214
Merck Index 10, 2822
Formula Weight 139.11
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Hydroxynicotinic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxynicotinic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'