missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethylfuran, Mikromol™

Product Code. 18093420
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Product Code. Quantity unitSize
18093420 100 mg 100mg
1 options
This item is not returnable. View return policy

Product Code. 18093420

Brand: Mikromol MM0877.310100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

2-Hydroxymethylfuran
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Furfuryl Alcohol
CAS 98-00-0
Analyte or Component Names Furfuryl Alcohol
Shipping Condition Room Temperature
Molecular Formula C5 H6 O2
Certifications/Compliance ISO 9001
Synonym 2-Hydroxymethylfuran, 2-Furylmethanol, 2-Furfurylalcohol
InChI Formula InChI=1S/C5H6O2/c6-4-5-2-1-3-7-5/h1-3,6H,4H2
SMILES OCc1occc1
IUPAC Name furan-2-ylmethanol
Molecular Weight (g/mol) 98.1
Quantity 100 mg
Formula Weight 98.0368 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.