missing translation for 'onlineSavingsMsg'
Learn More

2-(Hydroxymethyl)pyridine, 98%

Product Code. 10514071 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10514071

Brand: Thermo Scientific Acros 131991000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 586-98-1 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 586-98-1
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006348
InChI Key SHNUBALDGXWUJI-UHFFFAOYSA-N
Synonym 2-pyridinemethanol, 2-hydroxymethyl pyridine, piconol, 2-pyridinylmethanol, 2-pyridylmethanol, 2-pyridylcarbinol, pyridine-2-methanol, pyridine-2-carbinol, alpha-picolyl alcohol, pyridin-2-yl methanol
PubChem CID 11474
IUPAC Name pyridin-2-ylmethanol
SMILES C1=CC=NC(=C1)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 5.0°C
Color Brown or Colorless to Yellow
Density 1.1300g/mL
Boiling Point 220.0°C
Flash Point 117°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)CH2OH
Refractive Index 1.5410 to 1.5440
Quantity 100 g
Beilstein 21, I, 203
Specific Gravity 1.13
Solubility Information Solubility in water: soluble
Formula Weight 109.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-(Hydroxymethyl)pyridine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-(Hydroxymethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.