missing translation for 'onlineSavingsMsg'
Learn More

2-(Hydroxymethyl)pyridine-5-boronic acid, 97%

Product Code. 15420068
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 15420068

Brand: Thermo Scientific Alfa Aesar H53319.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 913835-98-0 | C6H8BNO3 | 152.944 g/mol

2-(Hydroxymethyl)pyridine-5-boronic acid is widely employed as intermediate in organic synthesis. It also acts as a laboratory reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Hydroxymethyl)pyridine-5-boronic acid is widely employed as intermediate in organic synthesis. It also acts as a laboratory reagent.

Notes
Store in dry, dark and ventilated place. Incompatible with strong oxidizi
TRUSTED_SUSTAINABILITY

Specifications

CAS 913835-98-0
Molecular Formula C6H8BNO3
MDL Number MFCD09027230
Quantity 250 mg
Synonym 6-hydroxymethyl pyridin-3-yl boronic acid, 6-hydroxymethyl pyridine-3-boronic acid, 2-hydroxymethyl pyridine-5-boronic acid, 6-hydroxymethyl pyridin-3-ylboronic acid, boronicacid, b-6-hydroxymethyl-3-pyridinyl, 5-boronopyridin-2-yl methanol, 6-hydroxymethyl-3-pyridineboronic acid, acmc-209raj, 6-hydroxymethyl-3-pyridyl boronic acid
InChI Key MSUQKXAJTUAESD-UHFFFAOYSA-N
SMILES B(C1=CN=C(C=C1)CO)(O)O
IUPAC Name [6-(hydroxymethyl)pyridin-3-yl]boronic acid
Molecular Weight (g/mol) 152.944
PubChem CID 44118255
Formula Weight 152.94
Percent Purity 97%
Chemical Name or Material 2-(Hydroxymethyl)pyridine-5-boronic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.