missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethyl Loratadine, TRC

Product Code. 30389237
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30389237 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30389237 Supplier Toronto Research Chemicals Supplier No. TRCH9477455MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxymethyl Loratadine
CAS 609806-39-5
Molecular Formula C23 H25 Cl N2 O3
Quantity 5 mg
Synonym 1-Piperidinecarboxylic acid, 4-[8-chloro-5,6-dihydro-2-(hydroxymethyl)-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-ylidene]-, ethyl ester, 2-Hydroxymethyl Loratadine
InChI Formula InChI=1S/C23H25ClN2O3/c1-2-29-23(28)26-11-9-15(10-12-26)21-20-8-6-18(24)13-17(20)4-3-16-5-7-19(14-27)25-22(16)21/h5-8,13,27H,2-4,9-12,14H2,1H3
SMILES CCOC(=O)N1CCC(=C2c3ccc(Cl)cc3CCc4ccc(CO)nc24)CC1
IUPAC Name ethyl 4-[8-chloro-2-(hydroxymethyl)-5,6-dihydrobenzo[1,2]cyclohepta[2,4-c]pyridin-11-ylidene]piperidine-1-carboxylate
Molecular Weight (g/mol) 412.91
Formula Weight 412.1554
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.