missing translation for 'onlineSavingsMsg'
Learn More

2-(Hydroxymethyl)benzeneboronic acid, 97%

Product Code. 15499177
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15499177

Brand: Thermo Scientific Alfa Aesar H52442.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 87199-14-2 | C7H9BO3 | 151.96 g/mol

2-(Hydroxymethyl)benzeneboronic acid participates in the reactivity of C-Terminal Cysteines with HNO.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Hydroxymethyl)benzeneboronic acid participates in the reactivity of C-Terminal Cysteines with HNO.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Keep cold.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87199-14-2
Molecular Formula C7H9BO3
Molecular Weight (g/mol) 151.96
MDL Number MFCD02258966
InChI Key AFHOBSCDNXGFMO-UHFFFAOYSA-N
Synonym 2-hydroxymethylphenylboronic acid, 2-hydroxymethyl phenylboronic acid, 2-hydroxymethyl phenyl boronic acid, 2-hydroxymethyl benzeneboronic acid, boronic acid, 2-hydroxymethyl phenyl, 2-hydroxymethylpheny, acmc-209qil, 2-hydromethylphenylboronicacid, ksc489m2p
PubChem CID 4374262
IUPAC Name [2-(hydroxymethyl)phenyl]boronic acid
SMILES OCC1=CC=CC=C1B(O)O

Specifications

Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 151.96
Percent Purity 97%
Chemical Name or Material 2-(Hydroxymethyl)benzeneboronic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.