missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethyl-5-nitrobenzeneboronic acid hemiester, 96%, Thermo Scientific™

Product Code. 15420486
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15420486 250 mg 250mg
15410486 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420486 Supplier Thermo Scientific Alfa Aesar Supplier No. H53212.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118803-40-0
Molecular Formula C7H6BNO4
Molecular Weight (g/mol) 178.938
MDL Number MFCD04115657
InChI Key GFFKBQCIGADRSN-UHFFFAOYSA-N
Synonym 6-nitrobenzo c 1,2 oxaborol-1 3h-ol, 1-hydroxy-6-nitro-2,1-benzoxaborolane, 2-hydroxymethyl-5-nitro benzeneboronic acid dehydrate, 6-nitro-3h-2,1-benzoxaborol-1-ol, 6-nitro-1,3-dihydro-2,1-benzoxaborol-1-ol, 2-hydroxymethyl-5-nitro phenylboronic acid, dehydrate, 2-hydroxymethyl-5-nitrophenylboronic acid, dehydrated, 6-nitro-2,1-benzoxaborol-1 3h-ol, 1,3-dihydro-1-hydroxy-6-nitro-2,1-benzoxaborole, 2,1-benzoxaborole,1,3-dihydro-1-hydroxy-6-nitro
PubChem CID 3813634
IUPAC Name 1-hydroxy-6-nitro-3H-2,1-benzoxaborole
SMILES B1(C2=C(CO1)C=CC(=C2)[N+](=O)[O-])O

Specifications

Quantity 250 mg
Formula Weight 178.94
Percent Purity 96%
Chemical Name or Material 2-Hydroxymethyl-5-nitrobenzeneboronic acid hemiester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.