missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethyl-18-crown-6, 97%

Product Code. 11470810
Change view
Click to view available options
Quantity:
0.25 g
1 g
Unit Size:
0.25g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470810 0.25 g 0.25g
11480810 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470810 Supplier Thermo Scientific Alfa Aesar Supplier No. 044477.01
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70069-04-4 | C13H26O7 | 294.34 g/mol

2-Hydroxymethyl-18-crown-6 is used for chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70069-04-4
Molecular Formula C13H26O7
Molecular Weight (g/mol) 294.34
MDL Number MFCD00188046
InChI Key HFRGASADQCZXHH-UHFFFAOYNA-N
Synonym 1,4,7,10,13,16-hexaoxacyclooctadecane-2-methanol, 2-hydroxymethyl-18-crown-6, 2-hydroxymethyl-18-crown 6-ether, 1,4,7,10,13,16-hexaoxacyclooctadecan-2-ylmethanol, hydroxy-18-crown-6, 18-crown-6-methanol, hydroxymethyl-18-crown-6, hfrgasadqczxhh-uhfffaoysa, 2-hydroxymethyl-18-crown, 2,5,8,11,14,17-hexaoxacyclooctadecyl methan-1-ol
PubChem CID 3611393
IUPAC Name 1,4,7,10,13,16-hexaoxacyclooctadec-2-ylmethanol
SMILES OCC1COCCOCCOCCOCCOCCO1

Specifications

Density 1.174 g/mL
Boiling Point 150°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.479
Quantity 0.25 g
Beilstein 4313301
Solubility Information Soluble in water (1000g/L) (25°C).
Formula Weight 294.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Hydroxymethyl-18-crown-6
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.