missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyisobutyric Acid, TRC

Product Code. 30011711
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011711 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30011711 Supplier Toronto Research Chemicals Supplier No. TRCH9429855G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 594-61-6
Molecular Formula C4H8O3
Quantity 5 g
Synonym α-Hydroxyisobutyric Acid, 2-Methyllactic Acid, 2-Hydroxy-2-methylpropanoic Acid, 2,2-Dimethylglycolic Acid, 2-Hydroxy-2-methylpropionic Acid, 2-Hydroxyisobutyric Acid, Acetonic Acid, Alpha-hydroxyisobutanoic Acid, Hydroxydimethylacetic Acid, NSC 402158, NSC 4505, α-HIB, α-Hydroxy-α-methylpropanoic Acid, α-Hydroxyisobutanoic Acid, α-Hydroxyisobutyric Acid;
InChI Formula InChI=1S/C4H8O3/c1-4(2,7)3(5)6/h7H,1-2H3,(H,5,6)
SMILES CC(C)(C(=O)O)O
IUPAC Name 2-hydroxy-2-methylpropanoic acid
Molecular Weight (g/mol) 104.1
Formula Weight 104.0473
Chemical Name or Material 2-Hydroxyisobutyric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.