missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyhippuric acid, 95%

Product Code. p-145366 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
This item is not returnable. View return policy

Product Code. 10061730

Brand: Thermo Scientific Acros 158620025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 487-54-7 | C9H9NO4 | 195.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 487-54-7
Molecular Formula C9H9NO4
Molecular Weight (g/mol) 195.17
MDL Number MFCD00002695
InChI Key ONJSZLXSECQROL-UHFFFAOYSA-N
Synonym salicyluric acid, 2-hydroxyhippuric acid, salicylurate, o-hydroxyhippuric acid, n-salicyloylglycine, salicyloylglycine, n-2-hydroxybenzoyl glycine, glycine, n-2-hydroxybenzoyl, 2-2-hydroxybenzamido acetic acid, n-o-hydroxybenzoylglycine
PubChem CID 10253
ChEBI CHEBI:9008
IUPAC Name 2-[(2-hydroxybenzoyl)amino]acetic acid
SMILES OC(=O)CNC(=O)C1=CC=CC=C1O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 164°C to 169°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Linear Formula HOC6H4CONHCH2CO2H
Quantity 2.5 g
Beilstein 10, 92
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol
Formula Weight 195.17
Percent Purity 95%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2-Hydroxyhippuric acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxyhippuric acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.