missing translation for 'onlineSavingsMsg'
Learn More

2'-Hydroxyflavone, TRC

Product Code. 30392463
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Product Code. Quantity unitSize
30392463 2.5 g 2.5g
1 options
This item is not returnable. View return policy

Product Code. 30392463

Brand: Toronto Research Chemicals TRCH9424782.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2'-Hydroxyflavone
CAS 35244-11-2
Molecular Formula C15 H10 O3
Quantity 2.5 g
Synonym 2-(2-Hydroxyphenyl)-4H-1-benzopyran-4-one, 2-(2-Hydroxyphenyl)-4H-chromen-4-one
InChI Formula InChI=1S/C15H10O3/c16-12-7-3-1-5-10(12)15-9-13(17)11-6-2-4-8-14(11)18-15/h1-9,16H
SMILES Oc1ccccc1C2=CC(=O)c3ccccc3O2
IUPAC Name 2-(2-hydroxyphenyl)chromen-4-one
Molecular Weight (g/mol) 238.238
Formula Weight 238.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.