missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethylflurazepam 0.1 mg/ml in Methanol, LoGiCal

Product Code. 30457484
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457484 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457484 Supplier LoGiCal Supplier No. LGCAMP0070.0702

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

2-Hydroxyethylflurazepam 0.1 mg/ml in Methanol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxyethylflurazepam
CAS 20971-53-3
Analyte or Component Names 2-Hydroxyethylflurazepam
Shipping Condition Room Temperature
Molecular Formula C17 H14 Cl F N2 O2
Certifications/Compliance ISO 17034
Synonym 2-Hydroxyethylflurazepam
InChI Formula InChI=1 S/C17H14ClFN2O2/c18-11-5-6-15-13(9-11)17(12-3-1-2-4-14(12)19)20-10-16(23)21(15)7-8-22/h1-6,9,22 H,7-8,10H2
SMILES OCCN1C(=O)CN=C(c2ccccc2F)c3cc(Cl)ccc13
IUPAC Name 7-chloro-5-(2-fluorophenyl)-1-(2-hydroxyethyl)-3 H-1,4-benzodiazepin-2-one
Solvent or Matrix Methanol
Industry Type Forensics
Molecular Weight (g/mol) 332.76
Quantity 1 mL
Formula Weight 332.0728 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.