missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl salicylate, 98+%, Thermo Scientific Chemicals

Product Code. 10147803 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10147803 50 mL Glass bottle 50mL
10229603 250 mL Glass bottle 250mL
10301434 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10147803 Supplier Thermo Scientific Acros Supplier No. 174100500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-28-5
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
MDL Number MFCD00002862
InChI Key LVYLCBNXHHHPSB-UHFFFAOYSA-N
Synonym 2-hydroxyethyl salicylate, glycol salicylate, ethylene glycol monosalicylate, espirosal, glycol monosalicylate, rheumacyl, sarocol, spirosal, glysal, phlogont
PubChem CID 6880
ChEBI CHEBI:86541
IUPAC Name 2-hydroxyethyl 2-hydroxybenzoate
SMILES OCCOC(=O)C1=CC=CC=C1O

Specifications

Melting Point 25°C
Color Colorless
Density 1.2500g/mL
Boiling Point 172°C (12.0 mmHg)
Flash Point 174°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula 2-(HO)C6H4CO2CH2CH2OH
Quantity 50 mL
Beilstein 10, 81
Merck Index 15, 4535
Specific Gravity 1.25
Solubility Information Solubility in water: 13g/L (20°C). Other solubilities: very soluble in alcohol,benzene,chloroform,,ether
Formula Weight 182.18
Percent Purity 98+%
Physical Form Oily Liquid or Low Melting Solid
Chemical Name or Material 2-Hydroxyethyl salicylate
Show More Show Less
compliance-icons
Product Identifier
  • 2-Hydroxyethyl salicylate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.