missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl stearate

Product Code. 10396831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
250 g
Packaging:
Glass bottle
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10396831 250 g Glass bottle 250g
10421173 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10396831 Supplier Thermo Scientific Acros Supplier No. 346952500
Outlet Corner
Use promo code 28080
View all promotions
 Restricted Item
In Stock
Add to basket
Edge
This item is not returnable. View return policy

CAS: 111-60-4 | C20H40O3 | 328.53 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-60-4
Molecular Formula C20H40O3
Molecular Weight (g/mol) 328.53
MDL Number MFCD00051465
InChI Key RFVNOJDQRGSOEL-UHFFFAOYSA-N
Synonym glycol stearate, cremophor a, 2-hydroxyethyl stearate, polyethylene glycol monostearate, ethylene glycol monostearate, cerasynt m, clearate g, cerasynt mn, cithrol ps, polyoxyl 40 stearate
PubChem CID 24762
IUPAC Name 2-hydroxyethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCCO

Specifications

Melting Point 55°C to 60°C
Color White to Yellow
Boiling Point 149°C
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 250 g
Solubility Information Solubility in water: insoluble in h2o
Formula Weight 328.53
Grade Technical
Concentration or Composition (by Analyte or Components) compound is a mixture of various chain-length esters and di(ethylene glycol) ester
Physical Form Flakes
Chemical Name or Material 2-Hydroxyethyl stearate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.