missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxyethyl Phosphate, TRC

Product Code. 30393515
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393515 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393515 Supplier Toronto Research Chemicals Supplier No. TRCH94205050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Hydroxyethyl Phosphate
CAS 1892-26-8
Molecular Formula C2H7O5P
Quantity 50 mg
Synonym 1-(Dihydrogen phosphate)1,2-Ethanediol, Ethylene glycol, Mono(dihydrogen phosphate), 2-Hydroxyethyl Dihydrogen Phosphate, 2-Hydroxyethyl Phosphate, Ethane-1,2-diol 1-Phosphate, Ethylene Glycol Monophosphate, Hydroxyethyl Phosphate, Mono(hydroxyethyl) Phosphate, Phosphoric Acid Mono(2-hydroxyethyl) Ester, β-Hydroxyethyl Phosphate
InChI Formula InChI=1S/C2H7O5P/c3-1-2-7-8(4,5)6/h3H,1-2H2,(H2,4,5,6)
SMILES OCCOP(=O)(O)O
Molecular Weight (g/mol) 142.05
Formula Weight 142.0031
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.