missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyethyl Phosphate-d4, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxyethyl Phosphate-d4 |
| Molecular Formula | C2H3D4O5P |
| Quantity | 25 mg |
| Synonym | 1-(Dihydrogen phosphate)1,2-Ethanediol-d4, Ethylene glycol, Mono(dihydrogen phosphate)-d4, 2-Hydroxyethyl Dihydrogen Phosphate-d4, 2-Hydroxyethyl Phosphate-d4, Ethane-1,2-diol 1-Phosphate, Ethylene Glycol Monophosphate-d4, Hydroxyethyl Phosphate-d4, Mono(hydroxyethyl) Phosphate-d4, Phosphoric Acid Mono(2-hydroxyethyl) Ester-d4, β-Hydroxyethyl Phosphate-d4 |
| InChI Formula | InChI=1S/C14H31O7P/c1-3-5-8-17-11-13-20-22(16,19-10-7-15)21-14-12-18-9-6-4-2/h15H,3-14H2,1-2H3/i7D2,10D2 |
| SMILES | OC([2H])([2H])C([2H])([2H])OP(O)(O)=O |
| Molecular Weight (g/mol) | 146.07 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?