missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Hydroxyethyl Methyl Terephthalate, TRC
Specifications
Specifications
| Chemical Name or Material | 2-Hydroxyethyl Methyl Terephthalate |
| CAS | 3645-00-9 |
| Molecular Formula | C11 H12 O5 |
| Quantity | 250 mg |
| Synonym | 1,4-Benzenedicarboxylic Acid 2-Hydroxyethyl Methyl Ester, Terephthalic Acid 2-Hydroxyethyl Methyl Ester, Ethylene Glycol Mono(methyl (Terephthalate), 2-Hydroxyethyl Methyl Terephthalate, Dimethyl Terephthalate-Ethylene Glycol Monoester, Methyl 2-Hydroxyethyl Terephthalate, Monoglycol Mnnomethyl Terephthalate, β-Hydroxyethyl Methyl Terephthalate |
| InChI Formula | InChI=1S/C11H12O5/c1-15-10(13)8-2-4-9(5-3-8)11(14)16-7-6-12/h2-5,12H,6-7H2,1H3 |
| SMILES | COC(=O)c1ccc(cc1)C(=O)OCCO |
| IUPAC Name | 1-O-(2-hydroxyethyl) 4-O-methyl benzene-1,4-dicarboxylate |
| Molecular Weight (g/mol) | 224.21 |
| Formula Weight | 224.0685 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?